Skip to Content
MilliporeSigma
All Photos(5)

Key Documents

WH0007453M2

Sigma-Aldrich

Monoclonal Anti-WARS antibody produced in mouse

clone 3A12, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-GAMMA2, Anti-IFI53, Anti-IFP53, Anti-tryptophanyl-tRNA synthetase

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

3A12, monoclonal

form

buffered aqueous solution

species reactivity

mouse, rat, human

technique(s)

immunoprecipitation (IP): suitable
indirect ELISA: suitable
indirect immunofluorescence: suitable
western blot: 1-5 μg/mL

isotype

IgG2aκ

GenBank accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... WARS(7453)

General description

Tryptophanyl-tRNA synthetase, cytoplasmic (WARS) or IFP53 is the cytoplasmic form of tryptophanyl-tRNA synthetase. It is induced by interferons and the gene encoding it is localized on human chromosome 14q32.31.

Immunogen

WARS (NP_004175, 50 a.a. ~ 149 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
YKAAAGEDYKADCPPGNPAPTSNHGPDATEAEEDFVDPWTVQTSSAKGIDYDKLIVRFGSSKIDKELINRIERATGQRPHHFLRRGIFFSHRDMNQVLDA

Biochem/physiol Actions

Tryptophanyl-tRNA synthetase, cytoplasmic (WARS) is involved in the tryptophan aminoacylation of tRNAtrp. Mutations in the gene encoding it have been associated with distal hereditary motor neuropathy.

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A recurrent WARS mutation is a novel cause of autosomal dominant distal hereditary motor neuropathy.
Tsai PC
Brain (2017)
Assignment of the human tryptophanyl-tRNA synthetase gene (WARS) to chromosome 14q32.2?q32.32
B?rglum A.D
Cytogenetic and genome research (1993)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service