Skip to Content
MilliporeSigma
All Photos(7)

Key Documents

WH0007157M1

Sigma-Aldrich

Monoclonal Anti-TP53 antibody produced in mouse

clone 2C3, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-LFS1, Anti-TRP53, Anti-p53, Anti-tumor protein p53 (Li-Fraumeni syndrome)

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

2C3, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
immunoprecipitation (IP): suitable
indirect ELISA: suitable
indirect immunofluorescence: suitable
western blot: 1-5 μg/mL

isotype

IgG1κ

GenBank accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TP53(7157)

General description

p53 is a tumor suppressor gene expressed in a wide variety of tissues. It is a tetrameric nuclear DNA-binding phosphoprotein. The gene encoding it is localized on human chromosome 17p13.1 and mouse chromosome 11.

Immunogen

TP53 (AAH03596, 94 a.a. ~ 201 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
SSSVPSQKTYQGSYGFRLGFLHSGTAKSVTCTYSPALNKMFCQLAKTCPVQLWVDSTPPPGTRVRAMAIYKQSQHMTEVVRRCPHHERCSDSDGLAPPQHLIRVEGNL

Biochem/physiol Actions

Tumor suppressor p53 has the capability to induce cell cycle arrest and has a role in DNA repair, senescence and apoptosis. It binds to Simian vacuolating virus 40 (SV40) T-antigen and human papilloma virus E6 protein. The p53 gene is mutated in various cancers, such as of the breast, ovarian, bladder, colon and lung.

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Multiple myeloma involving central nervous system: high frequency of chromosome 17p13.1 (p53) deletions
Hong Chang, et.al
British Journal of Haematology, 127(3), 280-284 (2004)
Cell Cycle and Apoptosis
Bruna Pucci, et.al
Neoplasia, 2(4), 291-299 (2000)
Multiple myeloma involving central nervous system: high frequency of chromosome 17p13.1 (p53) deletions.
Chang H
British Journal of Haematology, 127(3), 280-284 (2004)
Cell Cycle and Apoptosis
Bruna Pucci
Neoplasia, 2(4), 291-299 (2000)

Articles

p53 regulates gene expression, cell cycle control and functions as a tumor suppressor. Inactivation of p53 is closely tied to cancer development.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service