Skip to Content
MilliporeSigma
All Photos(4)

Key Documents

WH0005660M1

Sigma-Aldrich

Monoclonal Anti-PSAP antibody produced in mouse

clone 1D1-C12, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-FLJ00245, Anti-GLBA, Anti-MGC110993, Anti-SAP1, Anti-prosaposin, Anti-prosaposin (variant Gaucher disease and variant metachromatic leukodystrophy)

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

1D1-C12, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
immunoprecipitation (IP): suitable
indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG2aκ

GenBank accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PSAP(5660)

General description

Secreted prosaposin (PSAP) is a pleiotropic growth factor. It is found in bile, seminal plasma, human milk and cerebrospinal fluid. It also exists as a lysosomal protein. In lysosomes, PSAP is the precursor of four sphingolipid activator proteins (saposin A–D). The gene encoding PSAP is localized on human chromosome 10q22.1.

Immunogen

PSAP (AAH01503, 18 a.a. ~ 524 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
PVLGLKECTRGSAVWCQNVKTASDCGAVKHCLQTVWNKPTVKSLPCDICKDVVTAAGDMLKDNATEEEILVYLEKTCDWLPKPNMSASCKEIVDSYLPVILDIIKGEMSRPGEVCSALNLCESLQKHLAELNHQKQLESNKIPELDMTEVVAPFMANIPLLLYPQDGPRSKPQPKDNGDVCQDCIQMVTDIQTAVRTNSTFVQALVEHVKEECDRLGPGMADICKNYISQYSEIAIQMMMHMQPKEICALVGFCDEVKEMPMQTLVPAKVASKNVIPALELVEPIKKHEVPAKSDVYCEVCEFLVKEVTKLIDNNKTEKEILDAFDKMCSKLPKSLSEECQEVVDTYGSSILSILLEEVSPELVCSMLHLCSGTRLPALTVHVTQPKDGGFCEVCKKLVGYLDRNLEKNSTKQEILAALEKGCSFLPDPYQKQCDQFVAEYEPVLIEILVEVMDPSFVCLKIGACPSAHKPLLGTEKCIWGPSYWCQNTETAAQCNAVEHCKRHVWN

Biochem/physiol Actions

Prosaposin (PSAP) activates the androgen receptor and has been linked to stimulate metastasis in neoplasms. A variant in the PSAP gene leads to prosaposin deficiency which is a fatal infantile lysosomal storage disorder.

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Amplification and overexpression of prosaposin in prostate cancer
Shahriar Koochekpour
Public Health Nutrition null
Prosaposin, a regulator of estrogen receptor alpha, promotes breast cancer growth.
Wu Y
Cancer Science (2012)
Prosaposin activates the androgen receptor and potentiates resistance to endocrine treatment in breast cancer.
Breast Cancer Research (2015)
Clinical, biochemical and molecular characterization of prosaposin deficiency.
Motta M
Clinical Genetics (2016)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service