Skip to Content
MilliporeSigma
All Photos(6)

Key Documents

WH0005578M1

Sigma-Aldrich

Monoclonal Anti-PRKCA antibody produced in mouse

clone 2F11, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-AAG6, Anti-MGC129900, Anti-MGC129901, Anti-PKCA, Anti-PKCalpha, Anti-PRKACA, Anti-protein kinase C, alpha

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

2F11, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
indirect ELISA: suitable
indirect immunofluorescence: suitable
proximity ligation assay: suitable
western blot: 1-5 μg/mL

isotype

IgGκ

GenBank accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PRKCA(5578)

General description

Protein kinase C (PKC) is a family of serine- and threonine-specific protein kinases that can be activated by calcium and the second messenger diacylglycerol. PKC family members phosphorylate a wide variety of protein targets and are known to be involved in diverse cellular signaling pathways. PKC family members also serve as major receptors for phorbol esters, a class of tumor promoters. Each member of the PKC family has a specific expression profile and is believed to play a distinct role in cells. The protein encoded by this gene is one of the PKC family members. This kinase has been reported to play roles in many different cellular processes, such as cell adhesion, cell transformation, cell cycle checkpoint, and cell volume control. Knockout studies in mice suggest that this kinase may be a fundamental regulator of cardiac contractility and Ca(2+) handling in myocytes. (provided by RefSeq)

Immunogen

PRKCA (NP_002728, 563 a.a. ~ 672 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
KEAVSICKGLMTKHPAKRLGCGPEGERDVREHAFFRRIDWEKLENREIQPPFKPKVCGKGAENFDKFFTRGQPVLTPPDQLVIANIDQSDFEGFSYVNPQFVHPILQSAV

Biochem/physiol Actions

PRKCA (protein kinase C α) is a serine-threonine kinase. It is associated with tumorigenesis, invasion, apoptosis, differentiation, angiogenesis and metastasis. In endothelial cells, PRKCA-mediated phosphorylation of TRPC1 (transient receptor potential cation channel subfamily C member 1) regulates store operated Ca2+ entry. Downregulation of PKCA inhibits the invasion of urinary bladder, colon, renal cell, breast carcinomas, multiple myeloma, glioma and endometrial cancer cells.

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Inhibition of protein kinase Calpha prevents endothelial cell migration and vascular tube formation in vitro and myocardial neovascularization in vivo.
Wang A, et.al
Circulation Research, 90(5), 609-616 (2002)
PKCa promotes the mesenchymal to amoeboid transition and increases cancer cell invasiveness.
Vaskovicova K
BMC Cancer, 15 (2015)
Protein kinase Calpha phosphorylates the TRPC1 channel and regulates store-operated Ca2+ entry in endothelial cells.
Ahmmed GU, et.al
The Journal of Biological Chemistry, 279(20), 20941-20949 (2004)
Reduction of protein kinase C a (PKC-a) promote apoptosis via down-regulation of Dicer in bladder cancer.
Jiang Z, et.al
Journal of Cellular and Molecular Medicine, 19(5), 1085-1093 (2015)
MZF-1/Elk-1 Complex Binds to Protein Kinase Ca Promoter and Is Involved in Hepatocellular Carcinoma.
Yue CH, et.al
PLoS ONE, 10(5), e0127420-e0127420 (2015)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service