Skip to Content
MilliporeSigma
All Photos(4)

Key Documents

WH0005522M1

Sigma-Aldrich

Monoclonal Anti-PPP2R2C antibody produced in mouse

clone 6D1, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-IMYPNO, Anti-IMYPNO1, Anti-MGC33570, Anti-PR52, Anti-protein phosphatase 2 (formerly 2A), regulatory subunit B (PR 52), gamma isoform

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

6D1, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG1κ

GenBank accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... PPP2R2C(5522)

General description

PPP2R2C (protein phosphatase 2 regulatory subunit B γ) codes for a γ isoform of the subunit B55 subfamily. It is the regulatory subunit of protein phosphatase type 2A (PP2A). This gene is located on human chromosome 4p16.

Immunogen

PPP2R2C (NP_065149, 1 a.a. ~ 110 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
MGEDTDTRKINHSFLRDHSYVTEADIISTVEFNHTGELLATGDKGGRVVIFQREPESKNAPHSQGEYDVYSTFQSHEPEFDYLKSLEIEEKINKIKWLPQQNAAHSLLST

Biochem/physiol Actions

Absence of PPP2R2C (protein phosphatase 2 regulatory subunit B γ)
induces castration-resistance and is linked with increased prostate cancer-specific mortality. Suppressing PPP2R2C stimulates the development of prostate cancer in the absence of androgens without promoting androgen receptor (AR) activity in vitro. In mouse, PPP2R2C plays a major role in synaptic plasticity and hence learning and memory. It also acts as a tumor suppressor gene in human brain cancers.

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Over expression of PPP2R2C inhibits human glioma cells growth through the suppression of mTOR pathway.
Fan Y L, et al.
Febs Letters, 587(24), 3892-3897 (2013)
PPP2R2C, a gene disrupted in autosomal dominant intellectual disability.
Backx L, et al.
European Journal of Medical Genetics, 53(5), 239-243 (2010)
PPP2R2C loss promotes castration-resistant prostate cancer growth and is associated with increased prostate cancer-specific mortality.
Bluemn E G, et al.
Molecular Cancer Research (2013)
miR-1301 promotes prostate cancer proliferation through directly targeting PPP2R2C.
Bi D, et al.
Biomedicine and Pharmacotherapy, 81, 25-30 (2016)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service