Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

WH0004583M2

Sigma-Aldrich

Monoclonal Anti-MUC2 antibody produced in mouse

clone 1D10, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-MLP, Anti-mucin 2, intestinal/tracheal

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

1D10, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

indirect ELISA: suitable

isotype

IgG2aκ

GenBank accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MUC2(4583)

General description

The MUC2 (mucin 2, oligomeric mucus/gel-forming) protein belongs to the mucin protein family. It is an important mucin released in the large intestine. It is approximately a 550 kDa protein that has two PTS-domains, rich in proline, serine, and threonine. MUC2 is located on human chromosome 11p15.
This gene encodes a member of the mucin protein family. Mucins are high molecular weight glycoproteins produced by many epithelial tissues. The protein encoded by this gene is secreted and forms an insoluble mucous barrier that protects the gut lumen. The protein polymerizes into a gel of which 80% is composed of oligosaccharide side chains by weight. The protein features a central domain containing tandem repeats rich in threonine and proline that varies between 50 and 115 copies in different individuals. Alternatively spliced transcript variants of this gene have been described, but their full-length nature is not known. (provided by RefSeq)

Immunogen

MUC2 (NP_002448, 5081 a.a. ~ 5179 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
TTEVSYAGCTKTVLMNHCSGSCGTFVMYSAKAQALDHSCSCCKEEKTSQREVVLSCPNGGSLTHTYTHIESCQCQDTVCGLPTGTSRRARRSPRHLGSG*

Biochem/physiol Actions

Mucin 2, oligomeric mucus/gel-forming (MUC2) modulates the expression and antimicrobial activity of β-defensin 2. It acts as a major constituent of the two-layered mucous structure in the colon. Deficiency of MUC2 might lead to opportunistic microbial invasion and/or impaired innate responses.

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Colonic MUC2 mucin regulates the expression and antimicrobial activity of ?-defensin 2
Cobo ER, et al.
Mucosal Immunology, 8(6), 1360-1372 (2015)
Pooled Resequencing of 122 Ulcerative Colitis Genes in a Large Dutch Cohort Suggests Population-Specific Associations of Rare Variants in MUC2
Visschedijk MC, et al.
PLoS ONE (2016)
Transcriptional regulation of the 11p15 mucin genes. Towards new biological tools in human therapy, in inflammatory diseases and cancer
Van Seuningen I, et al.
Frontiers in Bioscience (2001)
Christian V Recktenwald et al.
The Journal of biological chemistry, 291(26), 13580-13590 (2016-04-30)
The main structural component of the mucus in the gastrointestinal tract is the MUC2 mucin. It forms large networks that in colon build the loose outer mucous layer that provides the habitat for the commensal flora and the inner mucous
Sjoerd van der Post et al.
Journal of proteome research, 13(12), 6013-6023 (2014-11-19)
The polymeric mucin MUC2 constitutes the main structural component of the mucus that covers the colon epithelium. The protein's central mucin domain is highly O-glycosylated and binds water to provide lubrication and prevent dehydration, binds bacteria, and separates the bacteria

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service