Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

WH0004359M5

Sigma-Aldrich

Monoclonal Anti-MPZ antibody produced in mouse

clone 3B12, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-CHM, Anti-CMT1, Anti-CMT1B, Anti-CMT2I, Anti-CMT2J, Anti-CMT4E, Anti-CMTDI3, Anti-DSS, Anti-HMSNIB, Anti-MPP, Anti-P0, Anti-myelin protein zero (Charcot-Marie-Tooth neuropathy 1B)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

3B12, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

indirect ELISA: suitable

isotype

IgG2aκ

GenBank accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... MPZ(4359)

General description

This gene encodes a major structural protein of peripheral myelin. Mutations in this gene result in the autosomal dominant form of Charcot-Marie-Tooth disease type 1 as other polyneuropathies. (provided by RefSeq)

Immunogen

MPZ (AAH06491.1, 1 a.a. ~ 258 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
MLRAPAPAPAMAPGAPSSSPSPILAVLLFSSLVLSPAQAIVVYTDREAHGAVGSRVTLHCSFWSSEWVSDDISFTWRYQPEGGRDAISIFHYAKGQPYIDEVGTFKERIQWVGDPRWKDGSIVIHNLDYSDNGTFTCDVKNPPDIVGKTSQVTLYVFEKVPTRYGVVLGAVIGGVLGVVLLLLLLFYVVRYCWLRRQAALQRRLSAMEKGKLHKPGKDASKRGRQTPVLYAMLDHSRSTKAVSEKKAKGLGESRKDKK

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service