Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

WH0001892M1

Sigma-Aldrich

Monoclonal Anti-ECHS1 antibody produced in mouse

clone 1G9, ascites fluid

Synonym(s):

Anti-SCEH, Anti-enoyl Coenzyme A hydratase, short chain, 1, mitochondrial

Sign Into View Organizational & Contract Pricing

Select a Size

200 μL
$406.00

$406.00


Ships within 2 weeks. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
200 μL
$406.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$406.00


Ships within 2 weeks. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

ascites fluid

antibody product type

primary antibodies

clone

1G9, monoclonal

species reactivity

human

technique(s)

immunoprecipitation (IP): suitable
indirect ELISA: suitable

isotype

IgMκ

GenBank accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ECHS1(1892)

Related Categories

Immunogen

ECHS1 (AAH08906, 13 a.a. ~ 290 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
GPLRPPVRCPAWRPFASGANFEYIIAEKRGKNNTVGLIQLNRPKALNALCDGLIDELNQALKIFEEDPAVGAIVLTGGDKAFAAGADIKEMQNLSFQDCYSSKFLKHWDHLTQVKKPVIAAVNGYAFGGGCELAMMCDIIYAGEKAQFAQPEILIGTIPGAGGTQRLTRAVGKSLAMEMVLTGDRISAQDAKQAGLVSKICPVETLVEEAIQCAEKIASNSKIVVAMAKESVNAAFEMTLTEGSKLEKKLFYSTFATDDRKEGMTAFVEKRKANFKDQ

Biochem/physiol Actions

ECHS1 (Enoyl CoA hydratase, short chain 1, mitochondrial) performs in the mitochondrial β-oxidation of fatty acid by catalyzing the second step of the process i.e. hydration of 2-trans-enoyl-coenzyme A (CoA) to L-3-hydroxyacyl-CoAs. It also participates in different metabolic pathways involving fatty acids and amino acids, valine. Study reports that heterozygous mutation in ECHS1 causes Leigh syndrome with hypotonia, metabolic acidosis, and developmental delay symptoms.

Physical form

Solution

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

U Janssen et al.
Genomics, 40(3), 470-475 (1997-03-15)
The second step in mitochondrial fatty acid beta-oxidation is catalyzed by short chain enoyl-CoA hydratase (ECHS1; EC 4.2.1.17). Inherited disorders of this pathway of energy metabolism present clinical and laboratory features resembling sudden infant death syndrome and Reye-like syndrome. To
M Kanazawa et al.
Enzyme & protein, 47(1), 9-13 (1993-01-01)
Short chain enoyl-CoA hydratase (SCEH) catalyzes the second step of the mitochondrial fatty acid beta-oxidation spiral. We isolated cDNA clones for human SCEH to facilitate investigation of the enzyme structure of the gene and to examine the genetic background of
Enzymes of fatty acid metabolism. II. Properties of crystalline crotonase.
J R STERN et al.
The Journal of biological chemistry, 218(2), 985-1002 (1956-02-01)
Chika Sakai et al.
Human mutation, 36(2), 232-239 (2014-11-14)
The human ECHS1 gene encodes the short-chain enoyl coenzyme A hydratase, the enzyme that catalyzes the second step of β-oxidation of fatty acids in the mitochondrial matrix. We report on a boy with ECHS1 deficiency who was diagnosed with Leigh

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service