Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

WH0001521M1

Sigma-Aldrich

Monoclonal Anti-CTSW antibody produced in mouse

clone 4A8, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-LYPN, Anti-cathepsin W (lymphopain)

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$541.00

$541.00


Ships within 2 weeks. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μG
$541.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$541.00


Ships within 2 weeks. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

4A8, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG2aκ

GenBank accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CTSW(1521)

General description

The protein encoded by this gene, a member of the peptidase C1 family, is a cysteine proteinase that may have a specific function in the mechanism or regulation of T-cell cytolytic activity. The encoded protein is found associated with the membrane inside the endoplasmic reticulum of natural killer and cytotoxic T-cells. Expression of this gene is up-regulated by interleukin-2. (provided by RefSeq)

Immunogen

CTSW (AAH48255, 22 a.a. ~ 376 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
IRGPLRAQDLGPQPLELKEAFKLFQIQFNRSYLSPEEHAHRLDIFAHNLAQAQRLQEEDLGTAEFGVTPFSDLTEEEFGQLYGYRRAAGGVPSMGREIRSEEPEESVPFSCDWRKVAGAISPIKDQKNCNCCWAMAAAGNIETLWRISFWDFVDVSVQELLDCGRCGDGCHGGFVWDAFITVLNNSGLASEKDYPFQGKVRAHRCHPKKYQKVAWIQDFIMLQNNEHRIAQYLATYGPITVTINMKPLQLYRKGVIKATPTTCDPQLVDHSVLLVGFGSVKSEEGIWAETVSSQSQPQPPHPTPYWILKNSWGAQWGEKGYFRLHRGSNTCGITKFPLTARVQKPDMKPRVSCPP

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thomas O Edinger et al.
mBio, 6(3), e00297-e00297 (2015-06-11)
Human cathepsin W (CtsW) is a cysteine protease, which was identified in a genome-wide RNA interference (RNAi) screen to be required for influenza A virus (IAV) replication. In this study, we show that reducing the levels of expression of CtsW

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service