Skip to Content
MilliporeSigma
All Photos(6)

Key Documents

WH0001497M9

Sigma-Aldrich

Monoclonal Anti-CTNS antibody produced in mouse

clone 5G6, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-CTNSLSB, Anti-PQLC4, Anti-cystinosis, nephropathic

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$541.00

$541.00


Ships within 2 weeks. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μG
$541.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$541.00


Ships within 2 weeks. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

5G6, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG2aκ

GenBank accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CTNS(1497)

Related Categories

General description

The cystinosin, lysosomal cystine transporter (CTNS) gene with 12 exons, spanning 23kb on genomic DNA, is mapped to human chromosome 17p13.3. The cystinosin protein contains seven-transmembrane-domains and is mainly localized in lysosomes.
This gene encodes a seven-transmembrane domain protein that functions to transport cystine out of lysosomes. Its activity is driven by the H+ electrochemical gradient of the lysosomal membrane. Mutations in this gene cause cystinosis, a lysosomal storage disorder. Alternative splicing results in multiple transcript variants. (provided by RefSeq)

Immunogen

CTNS (NP_004928, 1 a.a. ~ 100 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
MIRNWLTIFILFPLKLVEKCESSVSLTVPPVVKLENGSSTNVSLTLRPPLNATLVITFEITFRSKNITILELPDEVVVPPGVTNSSFQVTSQNVGQLTVY

Biochem/physiol Actions

Cystinosin (CTNS) plays a vital role in migrating cystine out of lysosomes. Mutations in CTNS gene is associated with the development of cystinosis, an autosomal recessive disease characterized by intralysosomal accumulation of cystine.

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Molecular analysis of the CTNS gene in Jordanian families with nephropathic cystinosisAnalisis molecular del gen CTNS en familias jordanas afectadas por cistinosis nefropatica
Jaradat S, et al.
Nefrologia : Publicacion Oficial de la Sociedad Espa?ola Nefrologia, 35, 547-553 (2015)
Impact of cystinosin glycosylation on protein stability by differential dynamic SILAC
Nevo N, et al.
Molecular and Cellular Proteomics, 16, 457-468 (2017)
First Report of CTNS Mutations in a Chinese Family with Infantile Cystinosis
Yang YJ, et al.
TheScientificWorldJournal (2015)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service