Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

WH0001231M1

Sigma-Aldrich

Monoclonal Anti-CCR2 antibody produced in mouse

clone 4D12, ascites fluid

Synonym(s):

Anti-CCCKR2, Anti-CCR2A, Anti-CCR2B, Anti-CKR2, Anti-CKR2A, Anti-CKR2B, Anti-CMKBR2, Anti-MCP1R, Anti-chemokine (C-C motif) receptor 2

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

ascites fluid

antibody product type

primary antibodies

clone

4D12, monoclonal

species reactivity

human

technique(s)

indirect ELISA: suitable
western blot: 1:500-1:1000

isotype

IgMκ

GenBank accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... CCR2(1231)

General description

C-C motif chemokine receptor 2 (CCR2) is encoded by the gene mapped to human chromosome 3p21.31. The encoded protein is a membrane-spanning G protein-coupled receptor for C-C motif chemokine ligand 2 (CCL2) as well as CCL7 and CCL13. CCR2 is a member of chemokine receptor subfamily of human class A G-protein-coupled receptors. The protein is expressed on monocytes, immature dendritic cells and T-cell subpopulations.

Immunogen

CCR2 (NP_000639, 1 a.a. ~ 42 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
MLSTSRSRFIRNTNESGEEVTTFFDYDYGAPCHKFDVKQIGA

Biochem/physiol Actions

C-C motif chemokine receptor 2 (CCR2) interacts with its ligand CCL2 and plays crucial roles in the muscle damage, repair and adaptation responses to eccentric exercise. The encoded protein is also implicated in maintenance of adipocyte function. In addition, it also regulates monocytes and macrophages trafficking, which are involved in development of atherosclerosis, obesity and type 2 diabetes. Mutations in the gene leads to exercise-induced muscle damage. Overexpression of CCR2 has been observed in nasopharyngeal carcinoma (NPC).

Physical form

Solution

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

CCR2 modulates inflammatory and metabolic effects of high-fat feeding
Weisberg SP, et al.
The Journal of Clinical Investigation, 116, 115-124 (2006)
CCL2-CCR2 axis promotes metastasis of nasopharyngeal carcinoma by activating ERK1/2-MMP2/9 pathway
Yang J, et al.
Oncotarget, 7, 15632-15647 (2016)
CCL2 and CCR2 polymorphisms are associated with markers of exercise-induced skeletal muscle damage
Hubal MJ, et al.
Journal of Applied Physiology, 108, 1651-1658 (2010)
Yi Zheng et al.
Nature, 540(7633), 458-461 (2016-12-08)
CC chemokine receptor 2 (CCR2) is one of 19 members of the chemokine receptor subfamily of human class A G-protein-coupled receptors. CCR2 is expressed on monocytes, immature dendritic cells, and T-cell subpopulations, and mediates their migration towards endogenous CC chemokine

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service