Skip to Content
MilliporeSigma
All Photos(7)

Key Documents

WH0001072M4

Sigma-Aldrich

Monoclonal Anti-CFL1 antibody produced in mouse

clone 1A1, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-CFL, Anti-cofilin 1 (non-muscle)

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

1A1, monoclonal

form

buffered aqueous solution

species reactivity

mouse, human, rat

technique(s)

indirect ELISA: suitable
indirect immunofluorescence: suitable
western blot: 1-5 μg/mL

isotype

IgG2aκ

GenBank accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CFL1(1072)

General description

Cofilin is a widely distributed intracellular actin-modulating protein that binds and depolymerizes filamentous F-actin and inhibits the polymerization of monomeric G-actin in a pH-dependent manner. It is involved in the translocation of actin-cofilin complex from cytoplasm to nucleus.
The cofilin 1 (CFL1) gene with four exons is mapped to human chromosome 11q13.1. The gene codes for an ubiquitously expressed, 18.5kDa phosphoprotein.

Immunogen

CFL1 (AAH11005, 1 a.a. ~ 166 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
MASGVAVSDGVIKVFNDMKVRKSSTPEEVKKRKKAVLFCLSEDKKNIILEEGKEILVGDVGQTVDDPYATFAKMLPDKDCRYALYDATYETKESKKEDLVFIFWAPESAPLKSKMIYASSKDAIKKKLTGIKHELQANCYEGVKDRCTLAEKLGGSAVISLEGKPL

Biochem/physiol Actions

Cofilin-l (CFL1) plays a vital role in the regulation of actin cytoskeleton reorganization. It is also implicated in cancer cell migration and invasion in several types of tumor tissues. Elevated expression of the gene is associated with the development of esophageal squamous cell carcinoma (ESCC). In addition, abnormal expression of the gene leads to vulvar cancer and thus, it can be considered as a potential therapeutic target for the same. CFL1 facilitates cytokinesis, cell motility and morphogenesis. Activated CFL1 helps in preventing cytoskeletal-associated defects in axonal development stimulated by synucleins (Syns).

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cofilin 1 activation prevents the defects in axon elongation and guidance induced by extracellular alpha-synuclein.
Tilve S
Scientific Reports (2015)
Association between CFL1 gene polymorphisms and spina bifida risk in a California population.
Zhu H
BMC Medical Genetics (2007)
Sreenath Ravindran et al.
Frontiers in molecular neuroscience, 12, 64-64 (2019-04-06)
Dendritic growth and branching are highly regulated processes and are essential for establishing proper neuronal connectivity. There is a critical phase of early dendrite development when these are heavily regulated by external cues such as trophic factors. Brain-derived neurotrophic factor
The role of cofilin-l in vulvar squamous cell carcinoma: A marker of carcinogenesis, progression and targeted therapy.
Wu Q
Oncology Reports, 35, 2743-2754 (2016)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service