Skip to Content
MilliporeSigma
All Photos(4)

Key Documents

WH0001045M1

Sigma-Aldrich

Monoclonal Anti-CDX2 antibody produced in mouse

clone 1C7, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-CDX3, Anti-caudal type homeo box transcription factor 2

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

1C7, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgGκ

GenBank accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CDX2(1045)

General description

Caudal-related homoeobox transcription factor 2 (CDX2) comprises a homeotic DNA-binding domain, transcriptional domain, and a regulatory domain. The CDX2 gene is mapped to human chromosome 13q12.2.
The level and beta-cell specificity of insulin gene expression are regulated by a set of nuclear proteins that bind to specific sequences within the promoter of the insulin gene (INS; MIM 176730) and interact with RNA polymerase to activate or repress transcription. The proteins LMX1 (MIM 600298) and CDX3 are homeodomain proteins that bind an A/T-rich sequence in the insulin promoter and stimulate its transcription (German et al., 1994 [PubMed 7698771]).[supplied by OMIM

Immunogen

CDX2 (AAH14461.1, 1 a.a. ~ 313 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
MYVSYLLDKDVSMYPSSVRHSGGLNLAPQNFVSPPQYPDYGGYHVAAAAAAAANLDSAQSPGPSWPAAYGAPLREDWNGYAPGGAAAAANAVAHGPNGGSPAAAMGYSSPADYHPHHHPHHHPHHPAAAPSCASGLLQTLNPGPPGPAATAAAEQLSPGGQRRNLCEWMRKPAQQSLGSQVKTRTKDKYRVVYTDHQRLELEKEFHYSRYITIRRKAELAATLGLSERQVKIWFQNRRAKERKINKKKLQQQQQQQPPQPPPPPPQPPQPQPGPLRSVPEPLSPVSSLQASVSGSVPGVLGPTGGVLNPTVTQ

Biochem/physiol Actions

Caudal-related homoeobox transcription factor 2 (CDX2) is very specific for intestinal mucosa maintenance and homeostasis. Along with β-catenin, CDX2 favors claudin-1 expression regulating tight junctions. This in turn is essential for the epithelial-mesenchymal transition (EMT) in colorectal cancer (CRC). Abnormal expression of CDX2 in the gut is implicated in intestinal-type metaplasias.

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jean-Noël Freund et al.
World journal of gastroenterology, 21(5), 1436-1443 (2015-02-11)
The homeoprotein encoded by the intestinal-specific Cdx2 gene is a major regulator of gut development and homeostasis, also involved in colon cancer as well as in intestinal-type metaplasias when it is abnormally expressed outside the gut. At the molecular level
Junhui Yu et al.
British journal of cancer, 124(1), 270-280 (2020-11-27)
Emerging evidence suggests the involvement of caudal-related homoeobox transcription factor 2 (CDX2) in tumorigenesis of various cancers. Although CDX2 functions in cancer invasion and metastasis, fewer studies focus on the role of CDX2 during the induction of epithelial-mesenchymal transition (EMT)
Svitlana V Malysheva et al.
Stem cell research, 30, 117-121 (2018-06-01)
Caudal-type homeobox 2 (CDX2) transcription factor is an important marker for early trophoblast lineages and intestinal epithelium. Due to its nuclear expression the immunostaining and sorting of viable CDX2pos cells is not possible. In this paper we report the generation
Johanne Davidsen et al.
BMC cancer, 20(1), 426-426 (2020-05-16)
Colon cancer is one of the most commonly diagnosed types of cancer with surgical resection of the tumor being the primary choice of treatment. However, the surgical stress response induced during treatment may be related to a higher risk of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service