Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

WH0000952M2

Sigma-Aldrich

Monoclonal Anti-CD38 antibody produced in mouse

clone 4G3, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-CD38 antigen (p45), Anti-T10

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

4G3, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

immunofluorescence: suitable
indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG1κ

GenBank accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... CD38(952)

General description

CD38 is a novel multifunctional ectoenzyme widely expressed in cells and tissues especially in leukocytes. CD38 also functions in cell adhesion,signal transduction and calcium signaling. (provided by RefSeq)
Cluster of differentiation 38 (CD38) is a glycosylated ∼45kDa type II transmembrane protein. It is encoded by the gene mapped to human chromosome 4p15.32.

Immunogen

CD38 (AAH07964, 201 a.a. ~ 300 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
CDVVHVMLNGSRSKIFDKNSTFGSVEVHNLQPEKVQTLEAWVIHGGREDSRDLCQDPTIKELESIISKRNIQFSCKNIYRPDKFLQCVKNPEDSSCTSEI

Biochem/physiol Actions

Cluster of differentiation 38 (CD38) plays a vital role in oxytocin secretion. In addition, it is also implicated in regulation of calcium concentration in airway smooth muscle. Aberrations in the gene expression is associated with the development of airway diseases like asthma. CD38 also functions in cell adhesion, signal transduction and calcium signaling. It serves as a potential disease marker for leukemias and myeloma.

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Deletion Involving CD 38 and BST 1 Results in a Fusion Transcript in a Patient With Autism and Asthma.
Ceroni F, et al.
Autism Research : Official Journal of the International Society for Autism Research, 7(2), 254-263 (2014)
CD38 at the junction between prognostic marker and therapeutic target.
Deaglio S, et al.
Trends in Molecular Medicine, 14(5), 210-218 (2008)
CD38/cADPR Signaling Pathway in Airway Disease: Regulatory Mechanisms.
Deshpande D A, et al.
Mediators of Inflammation, 2018 (2018)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service