Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

WH0000199M1

Sigma-Aldrich

Monoclonal Anti-AIF1 antibody produced in mouse

clone 2A2-B6, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-AIF1, Anti-IBA1, Anti-IRT1, Anti-allograft inflammatory factor 1

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

2A2-B6, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG1κ

GenBank accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... AIF1(199)

General description

This gene is induced by cytokines and interferon. Its protein product is thought to be involved in negative regulation of growth of vascular smooth muscle cells, which contributes to the anti-inflammatory response to vessel wall trauma. Three transcript variants encoding different isoforms have been found for this gene. (provided by RefSeq)

Immunogen

AIF1 (AAH09474.1, 1 a.a. ~ 147 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
MSQTRDLQGGKAFGLLKAQQEERLDEINKQFLDDPKYSSDEDLPSKLEGFKEKYMEFDLNGNGDIDIMSLKRMLEKLGVPKTHLELKKLIGEVSSGSGETFSYPDFLRMMLGKRSAILKVILMYEEKAREKEKPTGPPAKKAISELP

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Takehito Imamura et al.
European journal of obstetrics, gynecology, and reproductive biology, 180, 16-23 (2014-07-14)
Human heat shock protein 70 (HSP70) has been reported to enhance Toll-like receptor 4-mediated pelvic inflammation and growth of endometriotic cells. However, information on tissue expression of HSP70 before and after treatment with estrogen suppressing agent is scanty. Here, we
Anna Klinke et al.
American journal of respiratory cell and molecular biology, 51(1), 155-162 (2014-02-14)
Pulmonary arterial hypertension (PAH) is characterized by adverse remodeling of pulmonary arteries. Although the origin of the disease and its underlying pathophysiology remain incompletely understood, inflammation has been identified as a central mediator of disease progression. Oxidative inflammatory conditions support
Christoph Köhler
Cell and tissue research, 330(2), 291-302 (2007-09-18)
Ionized calcium-binding adapter molecule 1 (Iba1) is a 147-amino-acid calcium-binding protein widely in use as a marker for microglia. It has actin-crosslinking activity and is involved in aspects of motility-associated rearrangement of the actin cytoskeleton. The Iba1 gene and protein
Zhong-Ming Qian et al.
Molecular neurobiology, 50(3), 811-820 (2014-03-25)
Neuroinflammation is closely related to brain iron homeostasis. Our previous study demonstrated that lipopolysaccharides (LPS) can regulate expression of iron-regulatory peptide hepcidin; however, the mechanism is undefined. Here, we demonstrated that intracerebroventricular injection of LPS in rat brain upregulated hepcidin
Cindy X Guo et al.
Molecular vision, 20, 670-682 (2014-06-03)
Changes in connexin expression are associated with many pathological conditions seen in animal models and in humans. We hypothesized that gap junctions are important mediators in tissue dysfunction and injury processes in the retina, and therefore, we investigated the pattern

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service