Key Documents
W4261
Sigma-Aldrich
PCR Workstation
W × D × H 36 in. × 24 in. × 24 in., AC/DC input 120 V AC
About This Item
UNSPSC Code:
41100000
NACRES:
NB.22
availability
not available in EU
technique(s)
PCR: suitable
W × D × H
36 in. × 24 in. × 24 in.
General description
A self-contained work area that will help protect your PCR runs against contamination. Dual UV bulbs irradiate the work area prior to use, reducing the possibility that contaminating DNA will be amplified. Containment features reduce the chance of airborne contamination.
The non-ventilated hood has acrylic windows for a clear view, an acid-resistant black formica work surface, a laminated safety glass fascia, and double access doors. The germicidal lamps are controlled by a 24-hr timer. The work light is provided by dual fluorescent lamps, and there is a ceiling-mounted double AC outlet. Replacement part sold separately.
Legal Information
PCR Workstation is a trademark of C.B.S. Scientific Co.
Choose from one of the most recent versions:
Certificates of Analysis (COA)