W3766
Anti-WSTF antibody produced in rabbit
affinity isolated antibody, buffered aqueous solution
Synonym(s):
Anti-BAZ1B, Anti-Bromdomain Adjacent to Zn Finger Domain 1B, Anti-Williams Syndrome Transcription Factor, Anti-Williams-Beuren Syndrome Chromosome Region 9
About This Item
biological source
rabbit
conjugate
unconjugated
antibody form
affinity isolated antibody
antibody product type
primary antibodies
clone
polyclonal
form
buffered aqueous solution
mol wt
antigen 175 kDa
species reactivity
human
technique(s)
western blot: 1-2 μg/mL using lysates of HEK293-T cells
UniProt accession no.
shipped in
dry ice
storage temp.
−20°C
Gene Information
human ... BAZ1B(9031)
mouse ... Baz1b(22385)
rat ... Baz1b(368002)
Immunogen
Physical form
Not finding the right product?
Try our Product Selector Tool.
Storage Class
12 - Non Combustible Liquids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service