Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W2519

Sigma-Aldrich

Anti-WIPI-1 (N-terminal) antibody produced in rabbit

~1.0 mg/mL, affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-ATG18, Anti-WD repeat domain phosphoinositide interacting 1, Anti-WD40 repeat protein Interacting with phosphoInositides of 49kDa, Anti-WIPI-1α, Anti-WIPI49

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~49 kDa

species reactivity

human

concentration

~1.0 mg/mL

technique(s)

western blot: 2.5-5.0 μg/mL using whole extract of human G-361 cells.

UniProt accession no.

shipped in

dry ice

Storage temp.

−20°C

Gene Information

human ... WIPI1(55062)
mouse ... Wipi1(52639)
rat ... Wipi1(303630)

Immunogen

synthetic peptide corresponding to amino acids 46-61 of human WIPI-1, conjugated to KLH. The corresponding sequence is identical in mouse and rat.

Physical form

Solution in 0.01 M phosphate buffered saline pH 7.4, containing 15 mM sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service