Snake venom from Bitis caudalis (horned puff adder) may be used as a source of phospholipase A2/caudoxin and a wide variety of other other toxins such as dimeric disintegrins, PLA2 molecules, serine proteinases, a CRISP, C-type lectin-like proteins, L-amino acid oxidases, and snake venom metalloproteases. It may also be used as an immunogen.
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
Toxicon : official journal of the International Society on Toxinology, 24(4), 337-345 (1986-01-01)
Two toxic phospholipases A2, caudoxin from Bitis caudalis venom and the basic phospholipase A2 from Naja nigricollis venom, were selected for a study of the contribution of enzyme activity and charge properties to the presynaptic neurotoxicity and the contracture-inducing activity
Toxicon : official journal of the International Society on Toxinology, 20(3), 637-647 (1982-01-01)
Caudoxin, a single-chain phospholipase A2 isolated from the venom of Bitis caudalis is a toxic polypeptide with a formula weight of 13,332 dalton. The LD50 in mice (i.p.) was 0.18 (0.15-0.22) mg/kg. In the chick biventer cervicis muscle preparation the
Questions
Reviews
★★★★★ No rating value
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.