V7879
Anti-Vesicle-Associated Membrane Protein 1/2 antibody produced in sheep
IgG fraction of antiserum, buffered aqueous solution
Synonym(s):
Anti-VAMP 1/2
About This Item
biological source
sheep
Quality Level
conjugate
unconjugated
antibody form
IgG fraction of antiserum
antibody product type
primary antibodies
clone
polyclonal
form
buffered aqueous solution
mol wt
18 kDa
species reactivity
human
technique(s)
immunoprecipitation (IP): 1-5 μg/mL
western blot: 1-5 μg/mL
UniProt accession no.
shipped in
dry ice
storage temp.
−20°C
Gene Information
human ... VAMP1(6843) , VAMP2(6844)
Immunogen
Physical form
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class
10 - Combustible liquids
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service