Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V7009

Sigma-Aldrich

[Arg8]-Vasotocin-Gly-Lys

≥97% (HPLC)

Synonym(s):

Hydrin 1′

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C51H81N17O15S2
CAS Number:
Molecular Weight:
1236.42
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.26

Product Name

[Arg8]-Vasotocin-Gly-Lys, ≥97% (HPLC)

assay

≥97% (HPLC)

UniProt accession no.

storage temp.

−20°C

SMILES string

CC[C@H](C)[C@@H]1NC(=O)[C@H](Cc2ccc(O)cc2)NC(=O)[C@@H](N)CSSC[C@H](NC(=O)[C@H](CC(N)=O)NC(=O)[C@H](CCC(N)=O)NC1=O)C(=O)N3CCC[C@H]3C(=O)N[C@@H](CCCNC(N)=N)C(=O)NCC(=O)NCC(=O)N[C@@H](CCCCN)C(O)=O

InChI

1S/C51H81N17O15S2/c1-3-26(2)41-48(80)63-31(15-16-37(54)70)44(76)65-34(21-38(55)71)45(77)66-35(25-85-84-24-29(53)42(74)64-33(46(78)67-41)20-27-11-13-28(69)14-12-27)49(81)68-19-7-10-36(68)47(79)62-30(9-6-18-58-51(56)57)43(75)60-22-39(72)59-23-40(73)61-32(50(82)83)8-4-5-17-52/h11-14,26,29-36,41,69H,3-10,15-25,52-53H2,1-2H3,(H2,54,70)(H2,55,71)(H,59,72)(H,60,75)(H,61,73)(H,62,79)(H,63,80)(H,64,74)(H,65,76)(H,66,77)(H,67,78)(H,82,83)(H4,56,57,58)/t26-,29-,30-,31-,32-,33-,34-,35-,36-,41-/m0/s1

InChI key

YWRWVVYMJJBIRH-IWZJHKITSA-N

Gene Information

human ... AVP(551)

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Satomi Kohno et al.
General and comparative endocrinology, 132(3), 485-498 (2003-07-10)
In most amphibians, [Arg(8)] vasotocin (VT) has an antidiuretic effect that is coupled to the activation of adenylate cyclase. In contrast, mesotocin (MT) has a diuretic effect and acts via the inositol phosphate/calcium signaling pathway in amphibians. To further clarify
Y Ogushi et al.
The Journal of experimental biology, 213(2), 288-294 (2009-12-30)
The ventral pelvic skin of the tree frog Hyla japonica expresses two kinds of arginine vasotocin (AVT)-stimulated aquaporins (AQP-h2 and AQP-h3), which affect the capacity of the frog's skin to absorb water. As such, it can be used as a
Y Rouillé et al.
Proceedings of the National Academy of Sciences of the United States of America, 86(14), 5272-5275 (1989-07-01)
From neurointermediate pituitary glands of Xenopus laevis and Rana esculenta, previously unreported peptides termed hydrins, active on water permeability of frog urinary bladder and frog skin (Brunn or "water-balance" effect), have been isolated and sequenced. These peptides seem to be
S Iwamuro et al.
Biochimica et biophysica acta, 1176(1-2), 143-147 (1993-03-10)
The neurohypophysis of Xenopus and that of Ranidae and Bufonidae contain hydrin 1 (vasotocinyl-Gly-Lys-Arg) and hydrin 2 (vasotocinyl-Gly), respectively. In order to test the aldosterone-releasing activity of arginine vasotocin (AVT) and hydrin 1, purification of these peptides from an acid-extract
Yuji Ogushi et al.
American journal of physiology. Regulatory, integrative and comparative physiology, 299(5), R1150-R1162 (2010-09-03)
Regions of specialization for water absorption across the skin of Bufonid and Ranid anurans were identified by immunohistochemistry and Western blot analysis, using antibodies raised against arginine vasotocin (AVT)-stimulated aquaporins (AQPs) that are specific to absorbing regions of Hyla japonica.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service