Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V6505

Sigma-Aldrich

Valosin Porcine

≥97% (HPLC)

Synonym(s):

Peptide VQY, Val-Gln-Tyr-Pro-Val-Glu-His-Pro-Asp-Lys-Phe-Leu-Lys-Phe-Gly-Met-Thr-Pro-Ser-Lys-Gly-Val-Leu-Phe-Tyr

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C141H207N31O35S
CAS Number:
Molecular Weight:
2928.40
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.26

assay

≥97% (HPLC)

UniProt accession no.

storage temp.

−20°C

SMILES string

CSCCC(NC(=O)CNC(=O)C(Cc1ccccc1)NC(=O)C(CCCCN)NC(=O)C(CC(C)C)NC(=O)C(Cc2ccccc2)NC(=O)C(CCCCN)NC(=O)C(CC(O)=O)NC(=O)C3CCCN3C(=O)C(Cc4cnc[nH]4)NC(=O)C(CCC(O)=O)NC(=O)C(NC(=O)C5CCCN5C(=O)C(Cc6ccc(O)cc6)NC(=O)C(CCC(N)=O)NC(=O)C(N)C(C)C)C(C)C)C(=O)NC(C(C)O)C(=O)N7CCCC7C(=O)NC(CO)C(=O)NC(CCCCN)C(=O)NCC(=O)NC(C(C)C)C(=O)NC(CC(C)C)C(=O)NC(Cc8ccccc8)C(=O)NC(Cc9ccc(O)cc9)C(O)=O

InChI

1S/C141H207N31O35S/c1-77(2)63-97(157-128(193)100(66-84-33-18-14-19-34-84)159-122(187)93(39-24-27-58-144)154-130(195)102(71-114(182)183)161-132(197)107-40-28-59-170(107)139(204)104(70-88-72-147-76-150-88)164-124(189)95(52-54-113(180)181)156-137(202)117(81(9)10)168-134(199)109-42-29-60-171(109)138(203)103(68-86-43-47-89(175)48-44-86)163-123(188)94(51-53-110(145)177)155-135(200)115(146)79(5)6)126(191)153-92(38-23-26-57-143)121(186)158-99(65-83-31-16-13-17-32-83)120(185)149-73-111(178)151-96(55-62-208-12)125(190)169-118(82(11)174)140(205)172-61-30-41-108(172)133(198)166-106(75-173)131(196)152-91(37-22-25-56-142)119(184)148-74-112(179)167-116(80(7)8)136(201)162-98(64-78(3)4)127(192)160-101(67-85-35-20-15-21-36-85)129(194)165-105(141(206)207)69-87-45-49-90(176)50-46-87/h13-21,31-36,43-50,72,76-82,91-109,115-118,173-176H,22-30,37-42,51-71,73-75,142-144,146H2,1-12H3,(H2,145,177)(H,147,150)(H,148,184)(H,149,185)(H,151,178)(H,152,196)(H,153,191)(H,154,195)(H,155,200)(H,156,202)(H,157,193)(H,158,186)(H,159,187)(H,160,192)(H,161,197)(H,162,201)(H,163,188)(H,164,189)(H,165,194)(H,166,198)(H,167,179)(H,168,199)(H,169,190)(H,180,181)(H,182,183)(H,206,207)

InChI key

OVSVDLZVCMFSDM-UHFFFAOYSA-N

Gene Information

pig ... VCP(397524)

Biochem/physiol Actions

Stimulates pancreatic secretion.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

W E Schmidt et al.
FEBS letters, 191(2), 264-268 (1985-10-28)
The isolation and primary structure of a novel gastrointestinal peptide, designated valosin, is described. The peptide was purified from porcine upper gut extracts using an HPLC and N-terminal sequence screening strategy which depends on chromatographic and structural characteristics as isolation
Valosin stimulates gastric and exocrine pancreatic secretion and inhibits fasting small intestinal myoelectric activity in the dog.
Konturek SJ, Schmidt WE, et al.
Gastroenterologie Clinique Et Biologique, 92, 1181-1186 (1987)
Petra Hänzelmann et al.
The Journal of biological chemistry, 286(44), 38679-38690 (2011-09-15)
The AAA (ATPase associated with various cellular activities) ATPase p97, also referred to as valosin-containing protein (VCP), mediates essential cellular processes, including ubiquitin-dependent protein degradation, and has been linked to several human proteinopathies. p97 interacts with multiple cofactors via its

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service