Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V5752

Sigma-Aldrich

Val-Thr

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H18N2O4
CAS Number:
Molecular Weight:
218.25
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:

storage temp.

−20°C

SMILES string

CC(C)C(N)C(=O)NC(C(C)O)C(O)=O

InChI

1S/C9H18N2O4/c1-4(2)6(10)8(13)11-7(5(3)12)9(14)15/h4-7,12H,10H2,1-3H3,(H,11,13)(H,14,15)

InChI key

GVRKWABULJAONN-UHFFFAOYSA-N

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Studies on the conformation of amino acids. IX. Conformations of butyl, seryl, threonyl, cystennyl, and valyl residues in a dipeptide unit.
P K Ponnuswamy et al.
Biopolymers, 10(3), 565-582 (1971-01-01)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service