Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V5640

Sigma-Aldrich

Calcium-Like Peptide 3

Synonym(s):

CALP-3

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$248.00
25 MG
$833.00

$248.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$248.00
25 MG
$833.00

About This Item

UNSPSC Code:
51111800
NACRES:
NA.32

$248.00


Check Cart for Availability

Request a Bulk Order

form

lyophilized powder

Quality Level

color

white

solubility

water, double-distilled: soluble

storage temp.

−20°C

Amino Acid Sequence

H-Val-Lys-Phe-Gly-Val-Gly-Phe-Lys-OH

General description

The peptide activates the phosphodiesterase enzyme in the absence of calcium and inhibits cytotoxicity and apoptosis in a calcium mediated manner.

Physical form

Lyophilized from 0.1% aqueous TFA

Other Notes

Lyophilized from 0.1% TFA in H2O

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Calcium sensors as new therapeutic targets for airway hyperresponsiveness and asthma.
R Ten Broeke et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 15(10), 1831-1833 (2001-08-02)
M Villain et al.
The Journal of biological chemistry, 275(4), 2676-2685 (2000-01-25)
This report describes the use of the concept of inversion of hydropathy patterns to the de novo design of peptides targeted to a predetermined site on a protein. Eight- and 12-residue peptides were constructed with the EF hands or Ca(2+)-coordinating

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service