Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V5150

VPS34 human

recombinant, expressed in Sf9 cells

Synonym(s):

VPS34, N-terminal FLAG

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
Pricing and availability is not currently available.

recombinant

expressed in Sf9 cells

mol wt

98.2 kDa

storage temp.

−70°C

Looking for similar products? Visit Product Comparison Guide

General description

This Human VPS34 (vacuolar protein sorting 34) protein has a N-terminal FLAG-tag, expressed in Sf9 insect cells via a baculovirus expression system.

Application

Useful for the study of enzyme kinetics, screening inhibitors, and selectivity profiling.

Physical form

Formulated in a buffered solution consisting of 40 mM tris-HCl, pH 8.0, 110 mM NaCl, 2.2 mM KCl, 80 μg/ml FLAG peptide, and 20% glycerol.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service