Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V5140

Sigma-Aldrich

VPS34 human

recombinant, expressed in Sf9 cells

Synonym(s):

VPS34 human

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
$702.00

$702.00


Check Cart for Availability


Select a Size

Change View
20 μG
$702.00

About This Item

UNSPSC Code:
12352204
NACRES:
NA.54

$702.00


Check Cart for Availability

recombinant

expressed in Sf9 cells

Quality Level

form

liquid

specific activity

≥100 units/μg protein

mol wt

98.2 kDa

NCBI accession no.

shipped in

dry ice

storage temp.

−70°C

Looking for similar products? Visit Product Comparison Guide

General description

This Human VPS34 (vacuolar protein sorting 34) protein has a N-terminal FLAG-tag, expressed in Sf9 insect cells via a baculovirus expression system.

Application

Useful for the study of enzyme kinetics, screening inhibitors, and selectivity profiling.

Biochem/physiol Actions

VPS34 was shown to be required for proper liver and heart function. Mice deficient in VPS34 develop hepatomegaly and cardiomegaly. [1]

Unit Definition

One unit will transfer one picomole of phosphate from ATP to a phospholipid mixture per minute at 30 °C at pH 7.4.

Physical form

Formulatedin a buffered solution consisting of 40 mM Tris-HCl, pH 8.0, 110 mM NaCl, 2.2mM KCl, 80 μg/ml FLAG peptide, 20% glycerol., and 3 mM DTT.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nadia Jaber et al.
Proceedings of the National Academy of Sciences of the United States of America, 109(6), 2003-2008 (2012-02-07)
A critical regulator of autophagy is the Class III PI3K Vps34 (also called PIK3C3). Although Vps34 is known to play an essential role in autophagy in yeast, its role in mammals remains elusive. To elucidate the physiological function of Vps34

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service