Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V2390

Sigma-Aldrich

VRT-532

≥98% (HPLC)

Synonym(s):

4-Methyl-2-(5-phenyl-1H-pyrazol-3-yl)phenol, CFpot 532

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H14N2O
CAS Number:
Molecular Weight:
250.30
UNSPSC Code:
12352200

assay

≥98% (HPLC)

form

solid

solubility

DMSO: >20 mg/mL

storage temp.

−20°C

InChI

1S/C16H14N2O/c1-11-7-8-16(19)13(9-11)15-10-14(17-18-15)12-5-3-2-4-6-12/h2-10,19H,1H3,(H,17,18)

InChI key

AQWKVJFRGORALM-UHFFFAOYSA-N

Biochem/physiol Actions

VRT-532 is a mutated G551D CFTR corrector.
VRT-532 is a mutated G551D CFTR corrector. VRT-532 mediates G551D-CFTR by enhancement of ATP affinity of the mutant. VRT-532 also potentiates F508del mutated CFTR and works specifically on mutant CFTR rather than other membrane proteins.

Features and Benefits

This compound is featured on the Chloride Channels page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

pictograms

CorrosionExclamation mark

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1

Storage Class

11 - Combustible Solids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

We offer many products related to chloride channels for your research needs.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service