Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V1640

Sigma-Aldrich

VU0155041

≥98% (HPLC)

Synonym(s):

(±)-cis-2-(3,5-Dicholorphenylcarbamoyl)cyclohexanecarboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H15Cl2NO3
Molecular Weight:
316.18
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:

Quality Level

assay

≥98% (HPLC)

form

powder

color

white

solubility

DMSO: 54 mg/mL

SMILES string

OC(=O)[C@@H]1CCCC[C@@H]1C(=O)Nc2cc(Cl)cc(Cl)c2

InChI

1S/C14H15Cl2NO3/c15-8-5-9(16)7-10(6-8)17-13(18)11-3-1-2-4-12(11)14(19)20/h5-7,11-12H,1-4H2,(H,17,18)(H,19,20)/t11-,12+/m0/s1

InChI key

VSMUYYFJVFSVCA-NWDGAFQWSA-N

Biochem/physiol Actions

VU0155041 is a mixed allosteric agonist/positive allosteric modulator (PAM) of mGluR4.
VU0155041 is a mixed allosteric agonist/positive allosteric modulator (PAM) of mGluR4. VU0155041 is approximately 8-fold more potent than PHCCC and does not show any significant potentiator or antagonist activity at other mGluR subtypes. It is soluble in an aqueous vehicle and intracerebroventricular administration of 31-316 nmol of VU0155041 dose-dependently decreased haloperidol-induced catalepsy and reserpine-induced akinesia in rats. VU0155041 exhibits selectivity for mGluR4 relative to 67 different targets and does not affect the function of striatal NMDA receptors.
VU0155041 is a positive allosteric modulator of the metabotropic glutamate receptor subtype 4. It also shows some direct agonist activity, but at a site different from the glutamate binding site. /VU0155041 is approximately 8-fold more potent than PHCCC and enhances the activity of glutamate also about 8-fold. It shows promising anti-Parkinsonian effects in animal models of Parkinson′s disease.

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhichao Zhang et al.
Frontiers in molecular neuroscience, 13, 141-141 (2020-09-26)
Retinal progenitor cells (RPCs) remain in the eye throughout life and can be characterized by their ability for self-renewal as well as their specialization into different cell types. A recent study has suggested that metabotropic glutamate receptors (mGluRs) participate in
Zhichao Zhang et al.
Stem cells and development, 24(22), 2709-2722 (2015-07-16)
Promoting both endogenous and exogenous neural stem cells' (NSCs) survival in the hostile host environments is essential to cell replacement therapy for central nervous system (CNS) disorders. Type 4 metabotropic glutamate receptor (mGluR4), one of the members of mGluRs, has

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service