Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V1389

Sigma-Aldrich

Anti-Vesicle-Associated Membrane Protein 2 antibody produced in rabbit

affinity isolated antibody, lyophilized powder

Synonym(s):

Anti-Synaptobrevin 2, Anti-VAMP-2

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized powder

species reactivity

mouse, rat

technique(s)

immunohistochemistry: suitable using mouse brain sections
western blot (chemiluminescent): 0.3 μg/mL using rat brain whole protein extract

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... VAMP2(6844)
mouse ... Vamp2(22318)
rat ... Vamp2(24803)

Immunogen

peptide corresponding to residues 2-27 of mouse or rat VAMP-2. 25/26 residues identical in human.

Testing Method

Vendor Product Number: ANR-007 (Alomone)

Physical form

Lyophilized from a solution containing phosphate buffered saline, pH 7.4, 1% BSA, 5% sucrose, 0.025% NaN3.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Israr-Ul H Ansari et al.
Archives of biochemistry and biophysics, 618, 32-43 (2017-02-15)
Long-chain acyl-CoA synthetases (ACSLs) convert fatty acids to fatty acyl-CoAs to regulate various physiologic processes. We characterized the ACSL isoforms in a cell line of homogeneous rat beta cells (INS-1 832/13 cells) and human pancreatic islets. ACSL4 and ACSL3 proteins were
J R Woska et al.
Scandinavian journal of immunology, 73(1), 8-17 (2010-12-07)
Dysregulation of mast cell function contributes to allergic and autoimmune disease that affects more than 70 million persons in the United States alone. Identifying novel mast cell targets that mediate disease or disease progression is required for the development of
Yoosoo Yang et al.
The Biochemical journal, 450(3), 537-546 (2012-12-21)
Anti-allergic effects of dietary polyphenols were extensively studied in numerous allergic disease models, but the molecular mechanisms of anti-allergic effects by polyphenols remain poorly understood. In the present study, we show that the release of granular cargo molecules, contained in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service