Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V1260

Sigma-Aldrich

VEGF Receptor-3 (Flt-4)/Fc Chimera human

>95% (SDS-PAGE), recombinant, expressed in NSO cells, lyophilized powder

Synonym(s):

Vascular Endothelial Growth Factor Receptor‑3

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
51111800

biological source

human

recombinant

expressed in NSO cells

assay

>95% (SDS-PAGE)

form

lyophilized powder

mol wt

calculated mol wt ~112 kDa

packaging

pkg of 50 μg

storage condition

avoid repeated freeze/thaw cycles (Do not store in a frost-free freezer.)

impurities

endotoxin, tested

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... FLT4(2324)

Application

VEGF Receptor-3 (Flt-4)/Fc chimera human is a disulfide-linked homodimeric protein that consists of VEGF Receptor-3 fused with6X histidine tagged Fc portion of human IgG1 by a polypeptide linker. It may be used as a specific marker for lymphatic vessels as well as plays a pivotal role in the development of the lymphatic vessel system.

Biochem/physiol Actions

VEGF Receptor-3 (Flt-4) belongs to class III subfamily of RTKs and is expressed mainly in endothelial cells. VEGF Receptor-3 blocks VEGF-C/D and hence inhibits lymphangiogenesis and lymphatic metastasis in orthotopic urinary bladder cancer (OUBC).[1]

Physical form

Lyophilized from a 2 μm filtered solution in phosphate buffered saline.

Analysis Note

The binding affinity of recombinant VEGF R3 has not been determined. Its intended use is for western blot and ELISA standards.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Soluble vascular endothelial growth factor receptor-3 suppresses lymphangiogenesis and lymphatic metastasis in bladder cancer.
Molecular Cancer, 10 (2011)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service