Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V1139

Sigma-Aldrich

Anti-Vascular Endothelial Growth Factor Receptor 1 antibody produced in goat

affinity isolated antibody, lyophilized powder

Synonym(s):

Anti-Flt-1, Anti-VEGF R1

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
51111800

biological source

goat

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized powder

species reactivity

mouse

technique(s)

capture ELISA: 0.8 μg/mL
flow cytometry: 3-10 μg/mL
neutralization: suitable
western blot: 0.1-0.2 μg/mL

UniProt accession no.

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

mouse ... Flt1(14254)

General description

Vascular endothelial growth factors receptors (VEGF Rs) are receptor tyrosine kinases that bind to VEGF that plays important role in embryonic vasculogenesis, angiogenesis and homeostasis. The biological effects of VEGF are mediated by VEGF R1 and VEGF R2 that have the ability to heterodimerize. The binding of VEGF to VEGF R1 results in activation of tyrosine amino acids of the cytoplasmic tail of the receptor. This interaction results in the activation of MAPK, PI3K, PKC, FAK and Src kinase pathways. The biological roles of VEGF R1 include in epithelial cell growth, proliferation, survival, migration and vascular permeability and angiogenesis.
Anti-Vascular Endothelial Growth Factor Receptor-1 recognizes mouse VEGF R1. Based on ELISA, the antibody shows <7% cross reactivity with recombinant human VEGF R1, 4% cross-reactivity with recombinant mouse VEGF164, 1% cross-reactivity with recombinant rat VEGF164, and 0.2% cross-reactivity with recombinant mouse VEGF R2 and VEGF R3.

Specificity

The antibody will neutralize the bioactivity of recombinant mouse VEGF R1 and can be used in the blockade of receptor-ligand interaction.

Immunogen

purified recombinant mouse VEGF R1 extracellular domain expressed in NSO cells.

Application

Anti-Vascular Endothelial Growth Factor Receptor 1 antibody may be used in immunoblotting at a working concentration of 0.1-0.2 μg/ml. For flow cytometry the recommended concentration is 3-10 μg/ml. For ELISA, a working concentration of 0.8 μg/ml may be used. The antibody is suitable for neutralization reactions.

Physical form

Lyophilized from a 0.2 μm filtered solution in phosphate buffered saline with 5% trehalsoe.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Ananth Karumanchi et al.
Methods in molecular medicine, 122, 393-399 (2006-03-04)
Preeclampsia is characterized by the clinical triad of new hypertension, proteinuria, and edema after 20 wk of gestation. Recent evidence suggests that disturbances in angiogenic factors (such as vascular endothelial growth factor and placental growth factor) signaling and endothelial health
Hiroyuki Takahashi et al.
Clinical science (London, England : 1979), 109(3), 227-241 (2005-08-18)
The VEGF (vascular endothelial growth factor) family and its receptors are essential regulators of angiogenesis and vascular permeability. Currently, the VEGF family consists of VEGF-A, PlGF (placenta growth factor), VEGF-B, VEGF-C, VEGF-D, VEGF-E and snake venom VEGF. VEGF-A has at
Elizabeth Fleming et al.
BMC microbiology, 21(1), 278-278 (2021-10-16)
Genomics-driven discoveries of microbial species have provided extraordinary insights into the biodiversity of human microbiota. In addition, a significant portion of genetic variation between microbiota exists at the subspecies, or strain, level. High-resolution genomics to investigate species- and strain-level diversity
Masabumi Shibuya et al.
Experimental cell research, 312(5), 549-560 (2005-12-13)
The VEGF/VPF (vascular endothelial growth factor/vascular permeability factor) ligands and receptors are crucial regulators of vasculogenesis, angiogenesis, lymphangiogenesis and vascular permeability in vertebrates. VEGF-A, the prototype VEGF ligand, binds and activates two tyrosine kinase receptors: VEGFR1 (Flt-1) and VEGFR2 (KDR/Flk-1).
Anna-Karin Olsson et al.
Nature reviews. Molecular cell biology, 7(5), 359-371 (2006-04-25)
Vascular endothelial growth-factor receptors (VEGFRs) regulate the cardiovascular system. VEGFR1 is required for the recruitment of haematopoietic precursors and migration of monocytes and macrophages, whereas VEGFR2 and VEGFR3 are essential for the functions of vascular endothelial and lymphendothelial cells, respectively.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service