Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

UC289

Sigma-Aldrich

16β-Hydroxytestosterone

Synonym(s):

16β,17β-Dihydroxyandrost-4-en-3-one, 4-Androstene-16β,17β-diol-3-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H28O3
CAS Number:
Molecular Weight:
304.42
MDL number:
UNSPSC Code:
12161501
PubChem Substance ID:

form

powder

drug control

USDEA Schedule III; Home Office Schedule 4.2; regulated under CDSA - not available from Sigma-Aldrich Canada

color

white

mp

184-186

storage temp.

2-8°C

SMILES string

C[C@]12CC[C@H]3[C@@H](CCC4=CC(=O)CC[C@]34C)[C@@H]1C[C@H](O)[C@@H]2O

InChI

1S/C19H28O3/c1-18-7-5-12(20)9-11(18)3-4-13-14(18)6-8-19(2)15(13)10-16(21)17(19)22/h9,13-17,21-22H,3-8,10H2,1-2H3/t13-,14+,15+,16+,17+,18+,19+/m1/s1

InChI key

YMCWOAZGWMZGQT-KYQPOWKGSA-N

Application

16β-Hydroxytestosterone can be used as a standard for chromatographic assays of testosterone hydroxylases.

Biochem/physiol Actions

CYP3A2/2B1 (rat) metabolite; androgenic

pictograms

Health hazard

signalword

Danger

hcodes

Hazard Classifications

Carc. 2 - Repr. 1B

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Numazawa et al.
Analytical biochemistry, 146(1), 75-81 (1985-04-01)
A sensitive nonradiometric assay of aromatization of 16 alpha-hydroxylated androgens, 16 alpha-hydroxy-4-androstene-3,17-dione (16 alpha-OHA), and 16 alpha-hydroxytestosterone (16 alpha-OHT), has been developed using reverse-phase high-performance liquid chromatography with voltametric detector. The estrogens produced by human placental microsomes, estriol (E3) and
H B Lim et al.
Biochemistry international, 28(3), 543-550 (1992-11-01)
The metabolism of 16 alpha-hydroxytestosterone by cytochrome P-450 system was studied with two different inductions. At least 5 different metabolites with higher polarity were produced from 16 alpha-hydroxytestosterone. MT-B, one of their products, appeared to be specifically increased by 3-methylcholanthrene-
T L Klug et al.
Molecular genetics and metabolism, 89(4), 381-389 (2006-09-19)
Most, but not all, studies have found that women with a high urinary 2-hydroxyestrogen (2OHE) to 16alpha-hydroxyestrone (16alphaOHE1) ratio are at reduced risk for breast cancer and have a better prognosis. The aim was to identify factors associated with the
M Ringel et al.
Xenobiotica; the fate of foreign compounds in biological systems, 32(8), 653-666 (2002-09-26)
1. Steroids are known to act as permissive factors in hepatocytes. This study shows that dexamethasone (DEX) is a permissive factor for induction of CYP2B1/2, CYP3A1, CYP2A1 and probably also CYP2C11 in cultures with primary rat hepatocytes. 2. The induction
Paloma Riquelme et al.
Differentiation; research in biological diversity, 77(3), 263-276 (2009-03-11)
Under certain culture conditions human peripheral blood monocytes may be induced to express phenotypic markers of non-haematopoietic lineages, including hepatocyte-defining traits. One such example, the NeoHepatocyte, was previously shown to express a broad panel of hepatocyte-like marker antigens and metabolic

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service