Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

UC287

Sigma-Aldrich

7α-Hydroxytestosterone

Synonym(s):

4-Androstene-7α,17β-diol-3-one, 7α,17β-Dihydroxyandrost-4-en-3-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H28O3
Molecular Weight:
304.42
UNSPSC Code:
12161501
NACRES:
NA.77

form

solid

drug control

USDEA Schedule III; Home Office Schedule 4.2; regulated under CDSA - not available from Sigma-Aldrich Canada

color

white

mp

218-220 °C

storage temp.

2-8°C

Biochem/physiol Actions

7α-Hydroxytestosterone is known to block the synthesis of dihydrotestosterone and 5α-androstane-3α, 17β-diol in mature rat testis[1].
CYP2A1 (rat) metabolite; androgenic

pictograms

Health hazard

signalword

Danger

hcodes

Hazard Classifications

Carc. 2 - Repr. 1B

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P A Rosness et al.
Biochimica et biophysica acta, 488(1), 55-68 (1977-07-20)
1. Testicular 7 alpha-hydroxylation of testerone was assayed in cell extracts of rats between 12 and 79 days of age. Maximal 7 alpha-hydroxylase activity was observed about 60 days, while insignificant activity was obtained prior to 42 days of age.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service