Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

UC206

Sigma-Aldrich

Terfenadine alcohol metabolite

Synonym(s):

α-[4-(3-Hydroxy-2-methylprop-2-yl)phenyl]-4-(hydroxydiphenylmethyl)-1-piperidinebutanol

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$2,220.00

$2,220.00


Available to ship onApril 28, 2025Details


Request a Bulk Order

Select a Size

Change View
10 MG
$2,220.00

About This Item

Empirical Formula (Hill Notation):
C32H41NO3
Molecular Weight:
487.67
UNSPSC Code:
12161501
NACRES:
NA.77

$2,220.00


Available to ship onApril 28, 2025Details


Request a Bulk Order

form

solid

Quality Level

color

white

mp

123-128 °C

storage temp.

2-8°C

Application

Terfenadine alcohol metabolite can be used for metabolic assays of terfenadine.

Biochem/physiol Actions

CYP3A4 metabolitye of terfenadine

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Preparation Note

6-Hydroxywarfarin is soluble in DMSO.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service