Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

U8878

Sigma-Aldrich

Ultrogel® AcA 34

aqueous ethanol suspension, 60-140 μm (wet), exclusion limit 750,000 Da, fractionation range 20,000-350,000 Da (globular proteins)

Synonym(s):

Ultrogel® AcA

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
41104008

form

aqueous ethanol suspension

technique(s)

LPLC: suitable

matrix

3% acrylamide, 4% agarose

matrix active group

polymer

particle size

60-140 μm (wet)

pore size

20,000-350,000 Da fractionation range (globular proteins)
750,000 Da exclusion limit

pH

3-10

separation technique

size exclusion (SEC)

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Other Notes

Preswollen, composite polyacrylamide-agarose beads. The approximate % acrylamide and % agarose concentrations are indicated by the product name (Ultrogel Ac%A%).

Physical form

Suspension in 1 M NaCl and 20% ethanol

Legal Information

Ultrogel is a registered trademark of Pall Corporation

pictograms

Flame

signalword

Warning

hcodes

Hazard Classifications

Flam. Liq. 3

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

82.4 °F - closed cup

flash_point_c

28 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Mahapatra et al.
Fish physiology and biochemistry, 43(2), 477-491 (2017-03-02)
Two forms of vitellogenin (Vg: Vg1 and Vg2) were purified from the plasma of estradiol-17β (E

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service