Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

U8632

Sigma-Aldrich

Ubiquitin-Carrier Protein H5a, GST-tagged human

≥95% (SDS-PAGE), liquid

Synonym(s):

UbcH5a human

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352202

recombinant

expressed in E. coli

assay

≥95% (SDS-PAGE)

form

liquid

mol wt

47 kDa

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

General description

Manufactured for Sigma by Boston Biochem., Inc.

Biochem/physiol Actions

Human homologs of the yeast UBC4/5 family and play many important regulatory roles in inflammation and cancer.
Mediates the degradation of a myriad of short-lived regulatory proteins including the tumor suppressor p53 in the presence of E6/E6-AP or MDM2, the c-Fos transcription factor, and the signal-induced ubiquitination of IκBα, UBC5 is an essential gene in yeast.

Physical form

Solution in 50 mM HEPES, pH 8.0, 100 mM NaCl, 1 mM DTT in 10% glycerol.

Storage Class

12 - Non Combustible Liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The ubiquitin-proteasome proteolytic pathway.
A Ciechanover
Cell, 79(1), 13-21 (1994-10-07)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service