U8382
Ubiquitin Conjugating Enzyme Fractions mammalian
About This Item
biological source
rabbit
form
solution
technique(s)
ligand binding assay: suitable
shipped in
dry ice
storage temp.
−70°C
General description
Application
Biochem/physiol Actions
Storage Class
10 - Combustible liquids
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service