U7753
Uridine 5′-triphosphate-Agarose
matrix Cross-linked 4% beaded agarose, aqueous glycerol suspension
About This Item
Recommended Products
form
aqueous glycerol suspension
extent of labeling
1-3 μmol per mL
matrix
Cross-linked 4% beaded agarose
matrix activation
epoxy
matrix attachment
ribose hydroxyls
matrix spacer
22 atoms (adipic acid dihydrazide)
storage temp.
−20°C
Looking for similar products? Visit Product Comparison Guide
Application
Physical form
Disclaimer

Storage Class
12 - Non Combustible Liquids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service