Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

U6260

Sigma-Aldrich

Ubiquitin-Specific Protease 10 human

recombinant, expressed in Sf9 cells, ≥56% (SDS-PAGE)

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
$731.00

$731.00


Check Cart for Availability

Notify Me

Get notified when this item is ready to ship via email.


Select a Size

Change View
50 μG
$731.00

About This Item

UNSPSC Code:
12352204
NACRES:
NA.54

$731.00


Check Cart for Availability

Notify Me

Get notified when this item is ready to ship via email.

recombinant

expressed in Sf9 cells

Quality Level

form

buffered aqueous glycerol solution

mol wt

88 kDa

concentration

≥56% (SDS-PAGE)

NCBI accession no.

shipped in

dry ice

storage temp.

−70°C

General description

Human USP10 (Ubiquitin-Specific Protease 10) or Ubiquitin C-terminal Hydrolase 10, amino acids 2-798 (end) with an N-terminal FLAG-tag, expressed in a baculovirus-infected Sf9 cell expression system.

Application

Useful for the study of enzyme kinetics, screening inhibitors, and selectivity profiling.

Biochem/physiol Actions

Ubiquitin-specific protease 10 (USP10) expression has been shown to be induced by vasopressin. In HEK293 cells, USP10 was shown to deubiquitylate sorting nexin 3. [1]

Physical form

Solution in 40 mM Tris-HCl, 110 mM NaCl, 2.2 mM KCl, 80 ng/μl Flag peptide, 3 mM DTT, and 20% Glycerol at pH 8.0.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vasopressin-inducible ubiquitin-specific protease 10 increases ENaC cell surface expression by deubiquitylating and stabilizing sorting nexin 3.
Boulkroun S.
American Journal of Physiology: Renal Physiology, 295, F889-F900 (2008)
C Soncini et al.
Oncogene, 20(29), 3869-3879 (2001-07-06)
Degradation of cellular proteins through ubiquitination is a fundamental strategy for regulating biological pathways. De-ubiquitination, i.e. the removal of ubiquitin from proteins and peptides to which ubiquitin is attached, is catalyzed by processing proteases known as de-ubiquitinating enzymes. We are
Hortensia Faus et al.
Molecular and cellular endocrinology, 245(1-2), 138-146 (2005-12-22)
The role of the ubiquitin/proteasome system in degrading nuclear hormone receptors and regulating their transcriptional function has emerged in the last few years. We identified the ubiquitin-specific protease USP10 as part of DNA-bound androgen receptor (AR) complexes purified from nuclear
Jian Yuan et al.
Cell, 140(3), 384-396 (2010-01-26)
Stability and localization of p53 is essential for its tumor suppressor function. Ubiquitination by the E3 ubiquitin ligase Mdm2 is the major regulatory mechanism of p53, which induces p53 nuclear export and degradation. However, it is unclear whether ubiquitinated cytoplasmic

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service