U5760
(4R)-2-Undecyl-4-thiazolidinecarboxylic acid
≥98% (HPLC)
Synonym(s):
2-Undecylthiazolidine-4(R)-carboxylic acid
About This Item
assay
≥98% (HPLC)
form
powder
color
white to off-white
solubility
DMSO: >5 mg/mL
Storage temp.
2-8°C
SMILES string
CCCCCCCCCCCC1N[C@@H](CS1)C(O)=O
InChI
1S/C15H29NO2S/c1-2-3-4-5-6-7-8-9-10-11-14-16-13(12-19-14)15(17)18/h13-14,16H,2-12H2,1H3,(H,17,18)/t13-,14?/m0/s1
Inchi Key
FNBSOIBCKUUVJJ-LSLKUGRBSA-N
Biochem/physiol Actions
Features and Benefits
Preparation Note
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
Sigma-Aldrich offers many products related to lysophospholipid receptors for your research needs.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service