UBP302 is a selective GluR5 antagonist. UBP302 is the active enantiomer of UB296 and is one of the most potent GluR5 antagonists available for research use.
Preparation Note
UBP302 is soluble in DMSO at a concentration ≥5 mg/ml.
The excitatory neurotransmitter glutamate is essential in basal ganglia motor circuits and has long been thought to contribute to cell death and degeneration in Parkinson's disease (PD). While previous research has shown a significant role of NMDA and AMPA receptors
Toxicology and applied pharmacology, 284(2), 204-216 (2015-02-18)
Inhibition of acetylcholinesterase (AChE) after nerve agent exposure induces status epilepticus (SE), which causes brain damage or death. The development of countermeasures appropriate for the pediatric population requires testing of anticonvulsant treatments in immature animals. In the present study, exposure
The Journal of pharmacology and experimental therapeutics, 351(2), 359-372 (2014-08-27)
Exposure to nerve agents induces prolonged status epilepticus (SE), causing brain damage or death. Diazepam (DZP) is the current US Food and Drug Administration-approved drug for the cessation of nerve agent-induced SE. Here, we compared the efficacy of DZP with
Journal of biomolecular screening, 20(6), 708-719 (2015-02-24)
GluK1, a kainate subtype of ionotropic glutamate receptors, exhibits an expression pattern in the CNS consistent with involvement in pain processing and migraine. Antagonists of GluK1 have been shown to reduce pain signaling in the spinal cord and trigeminal nerve
We offers many products related to ion channel family glutamate receptors for your research needs.
Questions
Reviews
★★★★★ No rating value