Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

U21100-HH

Sigma-Aldrich

DMT-2′O-TBDMS-rU Phosphoramidite

Synonym(s):

5′-O-[bis(4-methoxyphenyl)phenylmethyl]-2′-O-[(1,1-dimethylethyl)dimethylsilyl]-uridine, 3′-[2-cyanoethyl N,N-bis(1-methylethyl)phosphoramidite], DMT-2′-O-TBDMS-rU amidite

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C45H61N4O9PSi
CAS Number:
Molecular Weight:
861.05
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

biological source

non-animal source (no BSE/TSE risk)

Quality Level

product line

Proligo Reagents

assay

≥99% (31P-NMR)
≥99.0% (reversed phase HPLC)

form

powder

packaging

pkg of 1 × 100 g U211011-01
pkg of 1 × 500 g U211051-01

technique(s)

oligo synthesis: suitable

impurities

≤0.3 wt. % water content (Karl Fischer)
≤0.5% P(III) Impurities 100-169ppm (31P-NMR)
≤0.5% single Impurity (reversed phase HPLC)
≤3 wt. % residual Solvent content

color

white to off-white

λ

conforms (UV/VIS Identity)

suitability

conforms to structure for H-NMR
conforms to structure for LC-MS

nucleoside profile

base: uridine
base protecting group: none
2' protecting group: TBDMS
5' protecting group: DMT
deprotection: standard

storage temp.

−20°C

SMILES string

COc1ccc(cc1)C(OC[C@H]2O[C@H]([C@H](O[Si](C)(C)C(C)(C)C)[C@@H]2OP(OCCC#N)N(C(C)C)C(C)C)N3C=CC(=O)NC3=O)(c4ccccc4)c5ccc(OC)cc5

InChI

1S/C45H61N4O9PSi/c1-31(2)49(32(3)4)59(55-29-15-27-46)57-40-38(56-42(48-28-26-39(50)47-43(48)51)41(40)58-60(10,11)44(5,6)7)30-54-45(33-16-13-12-14-17-33,34-18-22-36(52-8)23-19-34)35-20-24-37(53-9)25-21-35/h12-14,16-26,28,31-32,38,40-42H,15,29-30H2,1-11H3,(H,47,50,51)/t38-,40-,41-,42-,59?/m1/s1

InChI key

SKNLXHRBXYGJOC-ZMHKPELYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service