Key Documents
TRT51431
Multi Tasking Storage Cart
Hinged Door, yellow (drawers)
About This Item
UNSPSC Code:
12352200
NACRES:
JA.01
material
yellow (drawers)
manufacturer/tradename
TrippNT 51431
W × H × D
23 × 35 × 19 in.
weight
58 lb
General description
An Affordable Full Featured Lockable Polyethylene Organizing Cart with Drawers, Shelves and Dispensers
  • Organizes, stores and secures supplies
  • 3 large full extension drawers
  • Drawer: 12 3/4 x 3 5/8 x 15 7/8 WHD
  • Lockable sliding or hinged door
  • Bulk storage area
  • Document holder
  • Pen & tool holder
  • Gloves & Kimwipes® dispensers
  • Durable white polyethylene construction
  • 6 side shelves
  • 3 inch rubber casters (2 with brakes)
  • Built in, easy-to-grab handles
  • Professional, clean appearance
  • Fully assembled
Legal Information
Kimwipes is a registered trademark of Kimberly-Clark Worldwide, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)