Key Documents
TRT50440
Mobile Hazardous Waste Container
small
About This Item
UNSPSC Code:
12352200
NACRES:
JA.01
manufacturer/tradename
TrippNT 50440
W × H × D
15 × 24 × 15 in.
General description
  • Waste container for hazardous materials with visibility
  • 2 part construction allows lifting of primary container
  • 1/2 inch acrylic construction protects from beta emitters
  • 4 industrial casters; 2 with brakes
  • Hinged lid for easy filling, closing and containment
  • Standard hazard boxes fit these containers
  • Available in two sizes large and small
Choose from one of the most recent versions:
Certificates of Analysis (COA)