Key Documents
TMO340711
Nunc® cryogenic tubes
polypropylene, capacity 1.8 mL, round bottom, stepneck, external thread, sterile, case of 2000 ea
About This Item
UNSPSC Code:
41121704
material
polypropylene
round bottom
sterility
sterile
packaging
case of 2000 ea
manufacturer/tradename
Nalge-Nunc 340711
technique(s)
cryopreservation: suitable
capacity
1.8 mL
General description
Nunc®cryogenic tubes are designed for the storage of samples, ranging from general cold storage to the vapor phase of liquid nitrogen. These tubes are available with either internal or external threads to suit different preferences and requirements.
Legal Information
Nunc is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Choose from one of the most recent versions:
Certificates of Analysis (COA)