Key Documents
TMO167525
Nunc® Cell Factory(TM) filter adaptor cap
case of 100 ea
Synonym(s):
cell harvesting system, closed cell culture system
Select a Size
100 EACH
$54.50
$54.50
Check Cart for Availability
Select a Size
Change View
100 EACH
$54.50
About This Item
UNSPSC Code:
41161502
NACRES:
NB.24
$54.50
Check Cart for Availability
packaging
case of 100 ea
manufacturer/tradename
Nunc 167525
General description
Further the development of applications that require a more closed cell culture system using Thermo Scientific Nunc Cell Factory Accessories. Reduce the need for open interventions and support the use of aseptic methods for fluid handing relating to filling, inoculation, feeding and harvesting of the Cell Factory system.
Legal Information
Nunc is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Choose from one of the most recent versions:
Certificates of Analysis (COA)