Key Documents
T9692
Standard PCR microtubes
Strip tube (no cap)
About This Item
UNSPSC Code:
41121800
NACRES:
NB.22
material
natural
polypropylene tube
sterility
sterile
feature
RNase and DNase free
neck thread push fit
packaging
pkg of 120 strips
manufacturer/tradename
AB-0452
technique(s)
PCR: suitable
capacity
0.2 mL
working volume
0.2 mL
binding type
non-treated surface
General description
Precision moulded from virgin polypropylene and clean room produced.
Certified free from DNase, RNase and human genomic DNA contamination.
Thin-walled strips of 8 × 0.2 mL Thermo-Tubes
Choose from one of the most recent versions:
Certificates of Analysis (COA)