T9574
Anti-Tuberin (IA-22) antibody produced in rabbit
affinity isolated antibody, buffered aqueous solution
About This Item
biological source
rabbit
conjugate
unconjugated
antibody form
affinity isolated antibody
antibody product type
primary antibodies
clone
polyclonal
form
buffered aqueous solution
mol wt
antigen 180-200 kDa
species reactivity
rat, human, mouse
technique(s)
immunoprecipitation (IP): 5-10 μg using RIPA extract (0.5 mg) of human MCF-7 cells
western blot: 0.1-0.2 μg/mL using whole extracts of rat brain and mouse NIH3T3 cells
UniProt accession no.
shipped in
dry ice
storage temp.
−20°C
target post-translational modification
unmodified
Gene Information
human ... TSC1(7248)
mouse ... Tsc1(64930)
rat ... Tsc1(60445)
General description
Specificity
Immunogen
Application
- Chromatin immunoprecipitation (ChIP)
- electrophoretic mobility shift assay (EMSA)
- immunofluorescence staining
- immunoprecipitation
- immunoblotting
Biochem/physiol Actions
Physical form
Storage and Stability
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class
10 - Combustible liquids
wgk_germany
WGK 2
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service