T9288
Tygon® formula R-3603 laboratory tubing
I.D. × O.D. 3/8 in. × 5/8 in.
About This Item
Recommended Products
manufacturer/tradename
Tygon
I.D. × O.D.
3/8 in. × 5/8 in.
wall size
1/8 in.
General description
- Resists most inorganic chemicals
- Resistant to cracking and aging
- Low vapor pressure
- Lasts longer than rubber tubing
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)