T9038
Tygon® formula R-3603 laboratory tubing
I.D. × O.D. 3/4 in. × 1 in.
Synonym(s):
DEHP tubing alternative, chemical resistant tubing, laboratory tubing, vacuum tubing
All Photos(1)
About This Item
UNSPSC Code:
41105108
Recommended Products
manufacturer/tradename
Tygon
I.D. × O.D.
3/4 in. × 1 in.
wall size
1/8 in.
General description
Clear, general-purpose laboratory tubing. Ideal for peristaltic pumps, autoclavable to 248°F for 30 min. Temp. range: -49 to 158°F. Heavy walled Tygon is suitable for vacuum use.
- Resists most inorganic chemicals
- Resistant to cracking and aging
- Low vapor pressure
- Lasts longer than rubber tubing
Legal Information
Tygon is a registered trademark of Saint-Gobain Performance Plastics Corp.
Choose from one of the most recent versions:
Certificates of Analysis (COA)